Få mere at vide
Erdosteine, LoGiCal
Artikelnummer. 30463135
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463135 10 mg
1 options
Artikelnummer. 30463135 Leverandør LoGiCal Leverandørnr. LGCFOR0544.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Erdosteine
CAS 84611-23-4
Analyt- eller komponentnavne Erdosteine
Forsendelsestilstand Room Temperature
Molekylær formel C8 H11 N O4 S2
Certificeringer/compliance ISO 9001
Synonym Erdosteine;[[2-Oxo-2-[(tetrahydro-2-oxo-3-thienyl)amino]ethyl]thio]acetic acid, Dithiosteine, Erdostin, RV 144, Secresolv
InChI formel InChI=1S/C8H11NO4S2/c10-6(3-14-4-7(11)12)9-5-1-2-15-8(5)13/h5H,1-4H2,(H,9,10)(H,11,12)
Løsningstype Neat
SMIL OC(=O)CSCC(=O)NC1CCSC1=O
IUPAC navn 2-[2-oxo-2-[(2-oxothiolan-3-yl)amino]ethyl]sulfanylacetic acid
Industry Type Clinical Testing
Molekylvægt (g/mol) 249.31
Mængde 10 mg
Formel vægt 249.0129 g/mol