missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ ERCC8 Recombinant Rabbit Monoclonal Antibody (K01_2M32)

Artikelnummer. 30828826
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30828826 50 μL
30826359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30828826 Leverandør Invitrogen™ Leverandørnr. MA565518
 Begrænset vare
Forventet forsendelse: 22-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Rabbit Recombinant Monoclonal Antibody

ERCC8 Recombinant Monoclonal Antibody for Western Blot

This gene encodes a WD repeat protein, which interacts with Cockayne syndrome type B (CSB) protein and with p44 protein, a subunit of the RNA polymerase II transcription factor IIH. Mutations in this gene have been identified in patients with hereditary disease Cockayne syndrome (CS). CS cells are abnormally sensitive to ultraviolet radiation and are defective in the repair of transcriptionally active genes.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen ERCC8
Applikationer Western Blot
Klassifikation Recombinant Monoclonal
Klon K01_2M32
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene ERCC8
Genadgangsnr. Q13216
Gene Alias 2410022P04Rik; 2810431L23Rik; 4631412O06Rik; B130065P18Rik; Ckn1; Cockayne syndrome 1 (classical); Cockayne syndrome 1 homolog; cockayne syndrome WD repeat protein CSA; Cockayne syndrome WD repeat protein CSA homolog; Cockayne syndrome WD-repeat protein CSA; CSA; DNA excision repair protein ERCC-8; ERCC excision repair 8, CSA ubiquitin ligase complex subunit; Ercc8; excision repair cross-complementation group 8; excision repair cross-complementing rodent repair deficiency complementation group 8; excision repair cross-complementing rodent repair deficiency, complementation group 8; excision repaiross-complementing rodent repair deficiency, complementation group 8; UVSS2
Gensymboler ERCC8
Værtsarter Rabbit
Immunogen A synthetic peptide of human ERCC8.
Oprensningsmetode Protein A/G
Mængde 50 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 1161
Målarter Human
Indhold og opbevaring Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.