missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ ERC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597714
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
ERC1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
The protein encoded by this gene is a member of a family of RIM-binding proteins. RIMs are active zone proteins that regulate neurotransmitter release. This gene has been found fused to the receptor-type tyrosine kinase gene RET by gene rearrangement due to the translocation t(10;12)(q11;p13). Multiple transcript variants encoding different isoforms have been found for this gene.
Tekniske data
| ERC1 | |
| Polyclonal | |
| Unconjugated | |
| ERC1 | |
| 4930404L01Rik; 5033405M01Rik; 9630025C19Rik; B430107L16Rik; CAST; CAST2; CAZ-associated structural protein 2; Elks; ELKS/Rab6-interacting/CAST family member 1; ERC; ERC protein 1; Erc1; ERC-1; ERC1b; FLJ31750; KIAA1081; MGC12974; mKIAA1081; OTTHUMP00000160633; Rab6 interacting protein 2; RAB6-interacting protein 2; Rab6ip2; RAB6IP2A; RAB6IP2B; RIM-Binding Protein | |
| Rabbit | |
| Protein G | |
| RUO | |
| 23085 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 3.71 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8IUD2 | |
| ERC1 | |
| Recombinant Human ELKS/Rab6-interacting/CAST family member 1 protein (1-224AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion