missing translation for 'onlineSavingsMsg'
Learn More

Erbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7059199
Click to view available options
Quantity:
5 g
25 g
7440-52-0
Er
167.26
MFCD00010987
UYAHIZSMUZPPFV-UHFFFAOYSA-N
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
23980
CHEBI:33379
erbium
[Er]
7440-52-0
Er
167.26
MFCD00010987
UYAHIZSMUZPPFV-UHFFFAOYSA-N
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
23980
CHEBI:33379
erbium
[Er]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Color Gray
UN Number UN3089
Quantity 5 g
Percent Purity 99.9%
Mesh Size −40 Mesh
Sensitivity Air and moisture sensitive
Form Powder
Assay (REO)
Chemical Name or Material Erbium powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'