missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB4 (HER4)
ErbB4 (HER4)
Brand: Invitrogen PV3626
Additional Details : Weight : 0.05250kg
Description
Oncogene ERBB4 (HER4) is a member of the type I receptor protein tyrosine subfamily that includes EGFR, ERBB2, and ERBB3. ERBB4 binds and is activated by neuregulins and other factors and induces a variety of cellular responses including mitogenesis and differentiation.Specifications
NP_005226 | |
Oncology and Genetic Disease Research, Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
61.6kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
ErbB4 (HER4) | |
ERBB4 was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10 MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 708-993 | |
550nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL | |
GST Tag | |
Kinase and inhibitors | |
85% by SDS-PAGE |
0.34mg/mL | |
2066 | |
ERBB4 (HER4) | |
–80°C | |
1 Each | |
ErbB4 (HER4) | |
EGF receptor (ErbB) | |
ErbB4 (HER4) | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
For Research Use Only. Not for use in diagnostic procedures.