Få mere at vide
Eravacycline dihydrochloride, MedChemExpress
Artikelnummer. 16457238
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16457238 10 mg
16439767 5 mg
16403566 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16457238 Leverandør Medchem Express Leverandørnr. HY16980A
MedChemExpress Eravacycline dihydrochloride (TP-434 dihydrochloride) is a potent and broad-spectrum antibacterial agent.
Eravacycline dihydrochloride (TP-434 dihydrochloride) is a potent and broad-spectrum antibacterial agent.
Tekniske data
Kemisk navn eller materiale Eravacycline dihydrochloride
Fysisk form Powder
CAS 1334714-66-7
Farve Yellow
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C27H33Cl2FN4O8
Til brug med (applikation) COVID-19-immunoregulation
Synonym TP-434 dihydrochloride TP-434-046
Opløselighedsinformation DMSO : 220 mg/mL (348.39 mM; Need ultrasonic) ∣H2O : 50 mg/mL (79.18 mM; Need ultrasonic)
SMIL O=C(NC(C(O)=C1C2=O)=CC(F)=C1C[C@@]3([H])C[C@@]4([H])[C@H](N(C)C)C(O)=C(C(N)=O)C([C@@]4(O)C(O)=C32)=O)CN5CCCC5.[H]Cl.[H]Cl
Molekylvægt (g/mol) 631.48
Formel vægt 631.48
Procent renhed 96.93%