missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ER Beta Recombinant Protein, ligand binding domain, GST tag
ER Beta Recombinant Protein, ligand binding domain, GST tag
Brand: Invitrogen A15665
Additional Details : Weight : 0.01000kg
Description
This Estrogen Receptor (ER) Beta Ligand Binding Domain (LBD) is a 59.50 kDa recombinant human protein (amino acids 244-530) expressed as a GST fusion protein in baculovirus-infected insect cells. It was expressed and purified in the absence of exogenous ligands.ER Beta-LBD recombinant protein is:
ER Beta-LBD recombinant protein is manufactured under the strictest quality parameters. This product is guaranteed for 6 months after purchase. Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.
Used in:
LanthaScreen™ TR-FRET ER Beta Coactivator Assay Kit
For bulk reagents, email: !Mad-DiscoveryServices@lifetech.com
Specifications
NP_001428.1 | |
Nuclear Receptor Assay | |
ER Beta Recombinant Protein | |
–68 to –85°C | |
1 Each | |
ER Beta Recombinant Protein | |
aa 244 to 530 | |
Human | |
Insect |
1.24mg/mL | |
59.5kDa | |
25μg | |
For research use only. Not for use in diagnostic procedures | |
Estrogen receptor 2 (beta); oestrogen receptor; NR3A2 | |
0.9nM ER Beta - LBD (GST) is the final concentration recommended for the LanthaScreen™ TR-FRET ER Beta Competitive Binding Assay Goat (A15895). 7nM ER Beta - LBD (GST) is the final concentration recommended for the LanthaScreen™ TR-FRET ER Beta Coactivator Assay Goat (A15893) | |
ER Beta Recombinant Protein | |
GST Tag | |
98% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.