missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
ER beta/NR3A2 Antibody (ESR2/7006R), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-14205FR
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
ER beta/NR3A2 Monoclonal antibody specifically detects ER beta/NR3A2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Tekniske data
| ER beta/NR3A2 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| ESR2/7006R | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2 | |
| C-terminus fragment of recombinant human ER beta/NR3A2 protein (Uniprot: Q92731 ) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| 2100 | |
| Store at 4°C in the dark. | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion