Få mere at vide
Equilenin-d3, TRC
Artikelnummer. 30372744
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30372744 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30372744 Leverandør Toronto Research Chemicals Leverandørnr. TRCE5924025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Equilenin-d3
CAS 56588-54-6
Molekylær formel C18 2H3 H15 O2
Mængde 5 mg
Synonym Estra-1,3,5,7,9-pentaen-17-one-4,16,16-d3, 3-hydroxy- (9CI), 3-Hydroxyestra-1,3,5,7,9-pentaen-17-one-4,16,16-d3, Equilenin-d3, Equilenin-4,16,16-D3
InChI formel InChI=1S/C18H18O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h2-5,10,16,19H,6-9H2,1H3/t16-,18-/m0/s1/i7D2,10D
SMIL [2H]c1c(O)ccc2c3CC[C@@]4(C)[C@@H](CC([2H])([2H])C4=O)c3ccc12
IUPAC navn (13S,14S)-4,16,16-trideuterio-3-hydroxy-13-methyl-11,12,14,15-tetrahydrocyclopenta[a]phenanthren-17-one
Molekylvægt (g/mol) 269.35
Formel vægt 269.1495