missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Epsilon Dipeptide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Epsilon Dipeptide |
| CAS | 51513-74-7 |
| Molekylær formel | C31 H53 N5 O11 |
| Mængde | 100 mg |
| Synonym | L-Lysine, N6-[N2,N6-bis[(1,1-dimethylethoxy)carbonyl]-L-lysyl]-N2-[(1,1-dimethylethoxy)carbonyl]-, 2,5-pyrrolidinedione derivative |
| InChI formel | InChI=1S/C31H53N5O11/c1-29(2,3)44-26(41)33-19-13-10-14-20(34-27(42)45-30(4,5)6)24(39)32-18-12-11-15-21(35-28(43)46-31(7,8)9)25(40)47-36-22(37)16-17-23(36)38/h20-21H,10-19H2,1-9H3,(H,32,39)(H,33,41)(H,34,42)(H,35,43)/t20-,21-/m0/s1 |
| SMIL | CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O |
| IUPAC navn | (2,5-dioxopyrrolidin-1-yl) (2S)-6-[[(2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoyl]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate |
| Molekylvægt (g/mol) | 671.779 |
| Formel vægt | 671.374 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?