missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Eprosartan mesilat, Mikromol™
Description
Eprosartan mesilatat
Specifications
Specifications
| Kemisk navn eller materiale | Eprosartan Mesilate |
| CAS | 144143-96-4 |
| Analyt- eller komponentnavne | Eprosartan Mesilate |
| Forsendelsestilstand | Stuetemperatur |
| Molekylær formel | C23 H24 N2 O4 S . C H4 O3 S |
| Certificeringer/compliance | ISO 17034 |
| Synonym | 2-Thiophenepropanoic acid, α-[[2-butyl-1-[(4-carboxyphenyl)methyl]-1H-imidazol-5-yl]methylene]-, (E)-, monomethanesulfonate, 2-Thiophenepropanoic acid, α-[[2-butyl-1-[(4-carboxyphenyl)methyl]-1H-imidazol-5-yl]methylene]-, (αE)-, monomethanesulfonate, Eprosartan Mesylate, Eprosartan methanesulfonate, SKF 108566J |
| InChI formel | InChI=1S/C23H24N2O4S.CH4O3S/c1-2-3-6-21-24-14-19(12-18(23(28)29)13-20-5-4-11-30-20)25(21)15-16-7-9-17(10-8-16)22(26)27;1-5(2,3)4/h4-5,7-12,14H,2-3,6,13,15H2,1H3,(H,26,27)(H,28,29);1H3,(H,2,3,4)/b18-12+; |
| SMIL | CCCCc1ncc(\C=C(/Cc2cccs2)\C(=O)O)n1Cc3ccc(cc3)C(=O)O.CS(=O)(=O)O |
| IUPAC navn | 4-[[2-butyl-5-[(E)-2-carboxy-3-thiophen-2-ylprop-1-enyl]imidazol-1-yl]methyl]benzoesyre; metanesulfonsyre |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?