Få mere at vide
Eprosartan-d3, TRC
Artikelnummer. 30377845
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30377845 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377845 Leverandør Toronto Research Chemicals Leverandørnr. TRCE5901021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Eprosartan-d3
CAS 1185243-70-2
Molekylær formel C23H21D3N2O4S
Mængde 1 mg
Synonym (αE)-α-[[2-(Butyl-d3)-1-[(4-carboxyphenyl)methyl]-1H-imidazol-5-yl]methylene]-2-thiophenepropanoic Acid, (E)-2-(Butyl-d3)-1-(p-carboxybenzyl)-α-2-thenylimidazole- 5-acrylic Acid, Navixen-d3, SKB 108566-d3, SKF 108566-d3, Teveten-d3
InChI formel InChI=1S/C23H24N2O4S/c1-2-3-6-21-24-14-19(12-18(23(28)29)13-20-5-4-11-30-20)25(21)15-16-7-9-17(10-8-16)22(26)27/h4-5,7-12,14H,2-3,6,13,15H2,1H3,(H,26,27)(H,28,29)/b18-12+/i1D3
SMIL O=C(O)C1=CC=C(CN2C(/C=C(CC3=CC=CS3)/C(O)=O)=CN=C2CCCC([2H])([2H])[2H])C=C1
Molekylvægt (g/mol) 427.53