missing translation for 'onlineSavingsMsg'
Learn More

Koehler Technische Produkte™ Eppi Box

Brand:  Koehler Technische Produkte™ KEA50-C81NA

Additional Details : Weight : 0.12240kg

Product Code. 12414850

  • 121.00 DKK / Each
Estimated Shipment: 16-04-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Transparent
Cryo Box
9 x 9
130 mm
52 mm
Square
81 Vials Up to 12.5 mm Diameter
130 x 130 x 52 mm
130 mm
50 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers