Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
- Aerosol-tight options available.
Specifications
Specifications
| Type | Aerosol-tight Rotor |
| Capacity | 48 x 1.5 to 2.0 mL Tubes |
| Max. RCF | 18,213 x g |
| Max. Speed | 12,700 rpm |
| For Use With (Equipment) | Centrifuge 5427 R |
| Includes | Aerosol-tight with Rotor Lid |
| Model | FA-45-48-11 |