Learn More
Eppendorf™ A-4-38 Rotor Adapters
For round and rectangular buckets
Brand:  Eppendorf™ 5702733008
730.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15138354
  • 732.92 DKK / Pack of 2
Description
  • For use with Centrifuge 5702/ 5702 R/5702 RH and A-4-38 Rotor
  • Designed to meet the requirement of low- to mid-throughput clinical research and cell culture labs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor Adapter
1 x 5 mL Conical Tube
A-4-38
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Eppendorf™ A-4-38 Rotor Adapters > With 4 x 100mL round buckets