Få mere at vide
Tocris Bioscience™ Eplivanserin hemifumarate
GREENER_CHOICE
Potent and selective 5-HT2A antagonist
Brand:  Tocris Bioscience™ 4958/50
Encompass
Produktkode 15716356
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Eplivanserin hemifumarate is a potent and selective 5-HT2A antagonist (IC50 values are 5.8 and 120 and >100 nM for 5-HT2A, 5-HT2B and 5-HT2C, respectively). Attenuates cocaine-induced hyperactivity. Increases dopamine (DA) release in rat medial prefrontal cortex (mPFC); potentiates haloperidol-induced DA release in the mPFC and nucleus accumbens.
Tekniske data
Tekniske data
(1Z,2E)-1-(2-Fluorophenyl)-3-(4-hydroxyphenyl)-2-propen-1-one O-[2-(dimethylamino)ethyl]oxime hemifumarate
130580-02-8
SR-46349 hemifumarate, SR-46349B
50 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner