Få mere at vide
Eplerenone 7-Carboxylic Acid, TRC
Artikelnummer. 17932195
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17932195 100 mg
17970625 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17932195 Leverandør Toronto Research Chemicals Leverandørnr. TRCE588785100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 209253-82-7
Molekylær formel C23H28O6
Molekylvægt (g/mol) 400.46
Synonym Pregn-4-ene-7,21-dicarboxylic acid, 9,11-epoxy-17-hydroxy-3-oxo-, γ-lactone, (7α,11α,17α)-, (7α,11α,17α)-9,11-Epoxy-17-hydroxy-3-oxo-pregn-4-ene-7,21-dicarboxylic acid γ-lactone, 9,11α-Epoxy-7α-carboxy-3-oxo-17α-pregn-4-ene-21,17-carbolactone, 7α-Carboxy-9,11α-epoxy-3-oxo-17α-pregn-4-ene-21,17-carbolactone, (2'R)-9,11α-Epoxy-3,5'-dioxo-4',5'-dihydro-3'H-spiro[androst-4-ene-17,2'-furan]-7α-carboxylic acid, O-Demethyleplerenone, 9,11-Epoxy-17-hydroxy-3-oxo-pregn-4-ene-7,21-dicarboxylic Acid gamma-Lactone, Eplerenone EP Impurity D
IUPAC navn (1R,2S,9R,10R,11S,14R,15S,17R)-2,15-dimethyl-5,5'-dioxospiro[18-oxapentacyclo[8.8.0.01,17.02,7.011,15]octadec-6-ene-14,2'-oxolane]-9-carboxylic acid
SMIL C[C@]12C[C@H]3O[C@@]34[C@@H]([C@@H](CC5=CC(=O)CC[C@]45C)C(=O)O)[C@@H]1CC[C@@]26CCC(=O)O6
Tekniske data
Kemisk navn eller materiale Eplerenone 7-Carboxylic Acid
Mængde 100 mg
InChI formel InChI=1S/C23H28O6/c1-20-6-3-13(24)9-12(20)10-14(19(26)27)18-15-4-7-22(8-5-17(25)29-22)21(15,2)11-16-23(18,20)28-16/h9,14-16,18H,3-8,10-11H2,1-2H3,(H,26,27)/t14-,15+,16-,18+,20+,21+,22-,23-/m1/s1
Formel vægt 400.1886