Få mere at vide
Epicholesterol-2,2,3,4,4,6-d6, CDN
Artikelnummer. 30325349
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325349 0.01 g
30321845 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325349 Leverandør CDN Leverandørnr. CDND52350.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Epicholesterol-2,2,3,4,4,6-D6
CAS 92543-07-2
Molekylær formel C27 D6 H40 O
Mængde 0.01 g
Synonym Cholest-5-en-2,2,3,4,4,6-d6-3-ol, (3α)- (9 CI), (3α)-Cholest-5-en-2,2,3,4,4,6-d6-3-ol, (3α)-Cholest-5-en-3-ol-2,2,3,4,4,6-d6, [2,2,3,4,4,6-2H6]-Epicholesterol, Epicholesterol-2,2,3,4,4,6-D6, (3α)-Cholesterol-2,2,3,4,4,6-d6, (3α)-Cholesterol-d6, Epicholesterol-d6
InChI formel InChI=1 S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28 H,6-8,10-17H2,1-5H3/t19-,21-,22+,23-,24+,25+,26+,27-/m1/s1/i9D,13D2,17D2,21 D
SMIL [2 H]C1=C2C([2 H])([2 H])[C@]([2 H])(O)C([2 H])([2 H])C[C@]2(C)[C@H]3 CC[C@]4(C)[C@H](CC[C@H]4 [C@@H]3C1)[C@H](C)CCCC(C)C
IUPAC navn (3 R,8 S,9 S,10 R,13 R,14 S,17 R)-2,2,3,4,4,6-hexadeuterio-10,13-dimethyl-17-[(2 R)-6-methylheptan-2-yl]-1,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-ol
Molekylvægt (g/mol) 392.6905
Formel vægt 392.3925 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity