Få mere at vide
Epichlorohydrin-d5, TRC
Artikelnummer. 30372990
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30372990 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30372990 Leverandør Toronto Research Chemicals Leverandørnr. TRCE5823021G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Epichlorhydrin-d5
CAS 69533-54-6
Renhedsgrad noter GC
Molekylær formel C3 2H5 Cl O
Mængde 1 g
Synonym Epichlorhydrin-D5
InChI formel InChI=1S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2/i1D2,2D2,3D
SMIL [2H]C([2H])(Cl)C1([2H])OC1([2H])[2H]
IUPAC navn 2-[chloro(dideuterio)methyl]-2,3,3-trideuteriooxirane
Molekylvægt (g/mol) 97.56
Formel vægt 97.0343
Procent renhed >95