Få mere at vide
(±)-Epichlorohydrin-d5, CDN
Artikelnummer. 30311853
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30311853 1 g -
30309199 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311853 Leverandør CDN Leverandørnr. CDND11941G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Epichlorhydrin-d5
CAS 69533-54-6
Molekylær formel C3 2H5 Cl O
Mængde 1 g
Synonym Epichlorhydrin-D5
InChI formel InChI=1 S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2/i1D2,2D2,3 D
SMIL [2 H]C([2 H])(Cl)C1([2 H])OC1([2 H])[2 H]
IUPAC navn 2-[chloro(dideuterio)methyl]-2,3,3-trideuteriooxirane
Molekylvægt (g/mol) 97.56
Formel vægt 97.0343 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity