missing translation for 'onlineSavingsMsg'
Få mere at vide

epi-(-)-Strigolactone GR24, TRC

Artikelnummer. 30417711
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417711 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30417711 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6876151MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 188062-51-3
Molekylær formel C17H14O5
Molekylvægt (g/mol) 298.29
Synonym (3E,3aS,8bR)-3-[[[(2R)-2,5-Dihydro-4-methyl-5-oxo-2-furanyl]oxy]methylene]-3,3a,4,8b-tetrahydro-2H-indeno[1,2-b]furan-2-one, [3aS-[3E(S*),3aalpha,8balpha]]-3-[[(2,5-Dihydro-4-methyl-5-oxo-2-furanyl)oxy]methylene]-3,3a,4,8b-tetrahydro-2H-indeno[1,2-b]furan-2-one
IUPAC navn (3E,3aS,8bR)-3-[[(2R)-4-methyl-5-oxo-2H-furan-2-yl]oxymethylidene]-4,8b-dihydro-3aH-indeno[1,2-b]furan-2-one
SMIL CC1=C[C@H](O\C=C\2/[C@@H]3Cc4ccccc4[C@@H]3OC2=O)OC1=O

Specifications

Kemisk navn eller materiale epi-(-)-Strigolactone GR24
Mængde 1 mg
InChI formel InChI=1S/C17H14O5/c1-9-6-14(21-16(9)18)20-8-13-12-7-10-4-2-3-5-11(10)15(12)22-17(13)19/h2-6,8,12,14-15H,7H2,1H3/b13-8+/t12-,14+,15-/m0/s1
Formel vægt 298.0841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.