Få mere at vide
Epi Lovastatin-d3, TRC
Artikelnummer. 30379851
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30379851 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379851 Leverandør Toronto Research Chemicals Leverandørnr. TRCE58853210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Epi Lovastatin-d3
CAS 1330264-51-1
Molekylær formel C24H33D3O5
Mængde 10 mg
Synonym (2R)-2-(Methyl-d3)butanoic Acid (1S,3R,7S,8S,8aR)-1,2,3,7,8,8a-Hexahydro-3,7-dimethyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl Ester, Epilovastatin-d3
InChI formel InChI=1S/C24H36O5/c1-5-15(3)24(27)29-21-11-14(2)10-17-7-6-16(4)20(23(17)21)9-8-19-12-18(25)13-22(26)28-19/h6-7,10,14-16,18-21,23,25H,5,8-9,11-13H2,1-4H3/t14-,15+,16-,18+,19+,20-,21-,23-/m0/s1/i3D3
SMIL [2H]C([2H])([2H])[C@H](CC)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12
IUPAC navn [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] (2R)-2-(trideuteriomethyl)butanoate
Molekylvægt (g/mol) 407.56
Formel vægt 407.2751