missing translation for 'onlineSavingsMsg'
Få mere at vide

Epi Hydrocortisone 21-Acetate, TRC

Artikelnummer. 30136842
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136842 50 mg
17913365 25 mg
17951435 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30136842 Leverandør Toronto Research Chemicals Leverandørnr. TRCE58750050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1250-97-1
Molekylær formel C23 H32 O6
Molekylvægt (g/mol) 404.5
Synonym (11alpha)-21-(Acetyloxy)-11,17-dihydroxypregn-4-ene-3,20-dione, Pregn-4-ene-3,20-dione, 11alpha,17,21-trihydroxy-, 21-acetate (7CI,8CI), Progesterone, 11alpha,17,21-trihydroxy-, 21-acetate (6CI), 21-Acetoxy-11alpha,17-dihydroxypregn-4-ene-3,20-dione, Epihydrocortisone 21-acetate
IUPAC navn [2-[(8S,9S,10R,11R,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMIL CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@H](O)C[C@]12C

Specifications

Kemisk navn eller materiale Epi Hydrocortisone 21-Acetate
Mængde 50 mg
InChI formel InChI=1S/C23H32O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h10,16-18,20,26,28H,4-9,11-12H2,1-3H3/t16-,17-,18+,20+,21-,22-,23-/m0/s1
Formel vægt 404.2199
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.