Learn More
Invitrogen™ EPHA8 Recombinant Human Protein
EPHA8 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family.
Brand: Invitrogen™ PR7298B
Additional Details : Weight : 0.01000kg
Description
EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. EPHA8 plays a role in short-range contact-mediated axonal guidance during development of the mammalian nervous system.
Specifications
NP_065387 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
EPHA8, HEK6 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
76.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
EPH Receptor Kinase Family | |
Recombinant | |
Baculovirus |