Få mere at vide
EPA Method 525.2 Organochlorine Pesticide Mixture 500 μg/mL in Acetone, Dr. Ehrenstorfer
Artikelnummer. 30319840
Skift visning
Mængde:
1 ml
Artikelnummer. Mængde
30319840 1 ml
1 options
Artikelnummer. 30319840 Leverandør Dr. Ehrenstorfer Leverandørnr. DREGA09000336AC
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (+)-trans-Permethrin∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Chlorobenzilate∣Chloroneb∣Chlorothalonil∣Chlorthal-dimethyl∣Cis-Chlordane (Alpha Isomer)∣cis-Permethrin (mix of isomers)∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣Endrin-ketone∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Methoxychlor∣Trans-Chlordane (Gamma Isomer)∣trans-Nonachlor
CAS 51877-74-8, 72-54-8, 72-55-9, 50-29-3, 309-00-2, 959-98-8
Analyt- eller komponentnavne (+)-trans-Permethrin∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Chlorobenzilate∣Chloroneb∣Chlorothalonil∣Chlorthal-dimethyl∣Cis-Chlordane (Alpha Isomer)∣cis-Permethrin (mix of isomers)∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣Endrin-ketone∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Methoxychlor∣Trans-Chlordane (Gamma Isomer)∣trans-Nonachlor
Forsendelsestilstand Room Temperature
Certificeringer/compliance ISO 17034
Opløsningsmiddel eller Matrix Acetone
Mængde 1 ml
Koncentration eller sammensætning (efter analyt eller komponenter) (+)-trans-Permethrin 500 μg/mL∣4,4'-DDD 500 μg/mL∣4,4'-DDE 500 μg/mL∣4,4'-DDT 500 μg/mL∣Aldrin 500 μg/mL∣alpha-Endosulfan 500 μg/mL∣alpha-HCH 500 μg/mL∣beta-Endosulfan 500 μg/mL∣beta-HCH 500 μg/mL∣Chlorobenzilate 500 μg/mL∣Chloroneb 500 μg/mL∣Chlorothalonil 500 μg/mL∣Chlorthal-dimethyl 500 μg/mL∣Cis-Chlordane (Alpha Isomer) 500 μg/mL∣cis-Permethrin (mix of isomers) 500 μg/mL∣delta-HCH 500 μg/mL∣Dieldrin 500 μg/mL∣Endosulfan-sulfate 500 μg/mL∣Endrin 500 μg/mL∣Endrin-aldehyde 500 μg/mL∣Endrin-ketone 500 μg/mL∣gamma-HCH 500 μg/mL∣Heptachlor 500 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 500 μg/mL∣Methoxychlor 500 μg/mL∣Trans-Chlordane (Gamma Isomer) 500 μg/mL∣trans-Nonachlor 500 μg/mL