Learn More
Memmert™ Entry Port 23 mm for UN160, Standard Position Left Side Center/Center
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UN160F0
Product Code. 17559875
  • 959.00 DKK / Each
Only null left
Estimated Shipment: 25-07-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Entry port with 23 mm diameter
  • Flap and stopper closure
  • Standard position left side center/center
Specifications
Specifications
UN160 Oven
Entry Port
23 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Entry Port 23 mm for UN160, Standard Position Left Side Center/Center > For Use With: UN160