Learn More
Memmert™ Entry Port 23 mm for UN160, Custom TBC Right Side
Optional entry port for Memmert Ovens.
Brand: Memmert™ UN160F5
This item is not returnable.
View return policy
Description
- Entry port with 23 mm diameter
- Flap closure
- Custom position TBC in right side
Specifications
UN160 Oven | |
Entry Port | |
23 mm |
Product Content Correction
Product Title
Memmert™ Entry Port 23 mm for UN160, Custom TBC Right Side > For Use With: UN160
Spot an opportunity for improvement?Share a Content Correction