Learn More
Memmert™ Entry Port 23 mm for IN55, Standard Position Right Side Center/Center
Optional entry port for Memmert Incubators.
Brand: Memmert™ IN55F2
This item is not returnable.
View return policy
Description
- Entry port with 23 mm diameter
- Flap and stopper closure
- Standard position right side center/center
Specifications
IN55 Incubator | |
Entry Port | |
23 mm |
Product Content Correction
Product Title
Memmert™ Entry Port 23 mm for IN55, Standard Position Right Side Center/Center > For Use With: IN55
Spot an opportunity for improvement?Share a Content Correction