missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Enterovirus 71 VP2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5111997

Produktkode 17119863

  • 3445.00 DKK / 100µL

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

Enterovirus 71 VP2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Enterovirus 71 (EV71) causes hand-foot-and-mouth disease (HFMD), for which there are currently no therapeutics. EV71 virions have icosahedral symmetry and a diameter of approximately 30 nm, comprising 60 copies of the four protein subunits VP1 to VP4. Each of subunits VP1 to VP3 adopts a beta-barrel configuration common to many viruses.
TRUSTED_SUSTAINABILITY
Tekniske data

Tekniske data

Enterovirus 71 VP2
Polyclonal
Unconjugated
Enterovirus
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Enterovirus 71 VP2 (EV71 virus (strain USA/BrCr/1970)). The exact sequence is proprietary.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.64 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Rabbit
Antigen affinity chromatography
RUO
Virus
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.