missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Enterobacter aerogenes Antibody (57/82), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08869AF700
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Enterobacter aerogenes Monoclonal antibody specifically detects Enterobacter aerogenes in Bacteria samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Tekniske data
Enterobacter aerogenes | |
Monoclonal | |
Alexa Fluor 700 | |
Enterobacter aerogenes, Klebsiella mobilis | |
A crude sonicate of Enterobacter aerogenes | |
100 μg | |
Primary | |
Store at 4°C in the dark. | |
IgG κ |
ELISA, Immunofluorescence | |
57/82 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Bacteria | |
Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Enterobacter aerogenes Antibody (57/82), Alexa Fluor™ 700, Novus Biologicals™ > 100 μg; Alexa Fluor 700
Ser du en mulighed for forbedring?Del en indholdskorrektion