Få mere at vide
ent-Moxifloxacin Hydrochloride, TRC
Artikelnummer. 30136116
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136116 1 mg
17934745 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136116 Leverandør Toronto Research Chemicals Leverandørnr. TRCM7449901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346603-25-5
Molekylær formel C21H25ClFN3O4
Molekylvægt (g/mol) 437.89
Synonym 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aR,7aR)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic Acid Hydrochloride, d-Moxifloxacin Hydrochloride, (4R,7R)-Moxifloxacin Hydrochloride;
SMIL COC1=C2C(=CC(=C1N3C[C@@H]4CCCN[C@@H]4C3)F)C(=O)C(=CN2C5CC5)C(=O)O.Cl
Tekniske data
Kemisk navn eller materiale ent-Moxifloxacin Hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C21H24FN3O4.ClH/c1-29-20-17-13(19(26)14(21(27)28)9-25(17)12-4-5-12)7-15(22)18(20)24-8-11-3-2-6-23-16(11)10-24;/h7,9,11-12,16,23H,2-6,8,10H2,1H3,(H,27,28);1H/t11-,16+;/m0./s1