Få mere at vide
ent-Lamivudine Triphosphate Triethylammonium, TRC
Artikelnummer. 30475447
Skift visning
Mængde:
2 mg
Artikelnummer. Mængde
30475447 2 mg
1 options
Artikelnummer. 30475447 Leverandør Toronto Research Chemicals Leverandørnr. TRCL1732452MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale ent-Lamivudine Triphosphate Triethylammonium
CAS 2699949-69-2
Molekylær formel C8H14N3O12P3S . [C6H15N]x
Mængde 2 mg
Synonym ((2R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-1,3-oxathiolan-2-yl)methyl Tetrahydrogen Triphosphate Triethylammonium Salt
InChI formel InChI=1S/C8H14N3O12P3S.C6H15N/c9-5-1-2-11(8(12)10-5)6-4-27-7(21-6)3-20-25(16,17)23-26(18,19)22-24(13,14)15;1-4-7(5-2)6-3/h1-2,6-7H,3-4H2,(H,16,17)(H,18,19)(H2,9,10,12)(H2,13,14,15);4-6H2,1-3H3/t6-,7+;/m1./s1
SMIL CCN(CC)CC.NC1=NC(N([C@H]2CS[C@@H](COP(O)(OP(O)(OP(O)(O)=O)=O)=O)O2)C=C1)=O
IUPAC navn triethylamine ((2S,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-1,3-oxathiolan-2-yl)methyl triphosphate
Molekylvægt (g/mol) 570.4