Få mere at vide
ent-Lamivudine Acid (1S,2R,5S)-5-Methyl-2-isopropylcyclohexyl Ester, TRC
Artikelnummer. 30470236
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30470236 2.5 mg 2.5mg
30470235 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30470236 Leverandør Toronto Research Chemicals Leverandørnr. TRCL1724602.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale ent-Lamivudine Acid (1S,2R,5S)-5-Methyl-2-isopropylcyclohexyl Ester
CAS 147126-74-7
Molekylær formel C18H27N3O4S
Mængde 2.5 mg
Synonym (2S,5R)-5-(4-Amino-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolane-2-carboxylic Acid (1S,2R,5S)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester, [1S-[1alpha(2R*,5S*),2beta,5alpha]]-5-(4-Amino-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolane-2-carboxylic Acid 5-Methyl-2-(1-methylethyl)cyclohexyl Ester, ent-Lamivudine Acid (1S,2R,5S)-(+)-Menthol Ester, ent-Lamivudine Acid D-Menthol Ester
InChI formel InChI=1S/C18H27N3O4S/c1-10(2)12-5-4-11(3)8-13(12)24-16(22)17-25-15(9-26-17)21-7-6-14(19)20-18(21)23/h6-7,10-13,15,17H,4-5,8-9H2,1-3H3,(H2,19,20,23)/t11-,12+,13-,15+,17-/m0/s1
SMIL CC(C)[C@H]1CC[C@H](C)C[C@@H]1OC(=O)[C@H]2O[C@H](CS2)N3C=CC(=NC3=O)N
IUPAC navn [(1S,2R,5S)-5-methyl-2-propan-2-ylcyclohexyl] (2S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-1,3-oxathiolane-2-carboxylate
Molekylvægt (g/mol) 381.49
Formel vægt 381.1722