Få mere at vide
ent Atomoxetine-d3, Hydrochloride, TRC
Artikelnummer. 30335221
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30335221 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30335221 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7914041MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale ent Atomoxetine-d3 Hydrochloride
CAS 1217703-95-1
Molekylær formel C17H19D3ClNO
Mængde 1 mg
Synonym (γS)-N-Methyl-γ-(2-methyl-d3-phenoxy)benzenepropanamine Hydrochloride, (S)-(+)-Tomoxetine-d3 Hydrochloride, (S)-Tomoxetine-d3 Hydrochloride, LY 139602-d3
InChI formel InChI=1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H/t17-;/m0./s1/i2D3;
SMIL CC1=C(O[C@H](C2=CC=CC=C2)CCNC([2H])([2H])[2H])C=CC=C1.Cl
IUPAC navn (3S)-3-(2-methylphenoxy)-3-phenyl-N-(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 294.83