Få mere at vide
ent-3-Methoxymorphinan Hydrochloride, Mikromol™
Beskrivelse
ent-3-Methoxymorphinan Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Desmethyldextromethorphan Hydrochloride (ent-3-Methoxymorphinan Hydrochloride) |
| CAS | 1087-69-0 |
| Analyt- eller komponentnavne | N-Desmethyldextromethorphan Hydrochloride (ent-3-Methoxymorphinan Hydrochloride) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H23 N O . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Dextromethorphan Hydrobromide Imp. A (EP), Dextromethorphan Imp. A (EP), ent-3-Methoxymorphinan Hydrochloride, Dextromethorphan Hydrobromide Imp. A (EP) as Hydrochloride, N-Desmethyldextromethorphan Hydrochloride, Dextromethorphan Hydrobromide Impurity A as Hydrochloride, Dextromethorphan Impurity A as Hydrochloride |
| InChI formel | InChI=1S/C17H23NO.ClH/c1-19-13-6-5-12-10-16-14-4-2-3-7-17(14,8-9-18-16)15(12)11-13;/h5-6,11,14,16,18H,2-4,7-10H2,1H3;1H/t14-,16+,17+;/m1./s1 |
| SMIL | Cl.COc1ccc2C[C@@H]3NCC[C@]4(CCCC[C@H]34)c2c1 |
| IUPAC navn | (9α,13α,14α)-3-Methoxymorphinan hydrochloride |
| Vis mere |