Få mere at vide
ent-3-Methoxy-17-methylmorphinan-10-one, Mikromol™
Artikelnummer. p-200131098
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17456683 100 mg
17911056 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17456683 Leverandør Mikromol Leverandørnr. MM0033.02
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
ent-3-Methoxy-17-methylmorphinan-10-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale ent-3-Methoxy-17-methylmorphinan-10-one (10-Ketodextromethorphan)
CAS 57969-05-8
Analyt- eller komponentnavne ent-3-Methoxy-17-methylmorphinan-10-one (10-Ketodextromethorphan)
Forsendelsestilstand Room Temperature
Molekylær formel C18 H23 N O2
Certificeringer/compliance ISO 17025
Synonym ent-3-Methoxy-17-methylmorphinan-10-one, 10-Keto Dextromethorphan, Dextromethorphan Hydrobromide Imp. C (EP), Dextromethorphan Imp. C (EP), Dextromethorphan Related Compound C, Dextromethorphan Hydrobromide Impurity C, Dextromethorphan Impurity C
InChI formel InChI=1S/C18H23NO2/c1-19-10-9-18-8-4-3-5-14(18)16(19)17(20)13-7-6-12(21-2)11-15(13)18/h6-7,11,14,16H,3-5,8-10H2,1-2H3/t14-,16-,18+/m1/s1
SMIL COc1ccc2C(=O)[C@H]3[C@H]4CCCC[C@@]4(CCN3C)c2c1
IUPAC navn (9α,13α,14α)-3-Methoxy-17-methylmorphinan-10-one
Molekylvægt (g/mol) 285.38
Mængde 100 mg
Formel vægt 285.1729 g/mol