Få mere at vide
ent-17-Methylmorphinan-3-ol (L)-Tartrate, Mikromol™
Beskrivelse
ent-17-Methylmorphinan-3-ol (L)-Tartrate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dextrorphan tartrate |
| CAS | 143-98-6 |
| Analyt- eller komponentnavne | Dextrorphan tartrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H23 N O . C4 H6 O6 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | ent-17-Methylmorphinan-3-ol (L)-Tartrate, Dextromethorphan Hydrobromide Imp. B (EP) as (L)-Tartrate, Dextrorphan Tartrate, O-Desmethyldextromethorphan Tartrate, Dextromethorphan Hydrobromide Imp. B (EP), Dextromethorphan Imp. B (EP), Morphinan-3-ol, 17-methyl-, (9alpha,13alpha,14alpha)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1), Morphinan-3-ol, 17-methyl-, (9alpha,13alpha,14alpha)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1) (salt) (9CI), Morphinan-3-ol, 17-methyl-, (9alpha,13alpha,14alpha)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), Dextrorphan bitartrate, NIH 4591, d-3-Hydroxy-N-methylmorphinan tartrate, Dextromethorphan Hydrobromide Impurity B as (L)-Tartrate, Dextromethorphan Impurity B as (L)-Tartrate |
| InChI formel | InChI=1S/C17H23NO.C4H6O6/c1-18-9-8-17-7-3-2-4-14(17)16(18)10-12-5-6-13(19)11-15(12)17;5-1(3(7)8)2(6)4(9)10/h5-6,11,14,16,19H,2-4,7-10H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t14-,16+,17+;1-,2-/m11/s1 |
| SMIL | CN1CC[C@@]23CCCC[C@@H]2[C@@H]1Cc4ccc(O)cc34.O[C@H]([C@@H](O)C(=O)O)C(=O)O |
| IUPAC navn | (9α,13α,14α)-3-Hydroxy-17-methylmorphinan (2R,3R)-2,3-dihydroxybutanoate |
| Vis mere |