missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENPP-5 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99809UV
Additional Details : Weight : 0.00970kg
Description
ENPP-5 Polyclonal specifically detects ENPP-5 in Human samples. It is validated for ELISA.Specifications
ENPP-5 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
59084 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 350 | |
EC 3.1, ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative function), ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative), ectonucleotide pyrophosphatase/phosphodiesterase family member 5, E-NPP 5, KIAA0879, NPP-5 | |
Produced in rabbits immunized with purified, recombinant ENPP-5 (Q9UJA9; Met1-Gly429). | |
0.1 mL | |
Primary | |
Human | |
Purified |