Learn More
Ark-Plas Products™ Enlarging Check Valve
Created from Kynar. Ark-Plas Products™ Enlarging Check Valve is excellent for prevention of backflow of fluid. The valve allows for an increase in pressure of the fluid.
Brand:  Ark-Plas Products™ HDX59-DF0
Additional Details : Weight : 0.04590kg
 View more versions of this product
Product Code. 11841180
  • 1172.60 DKK / Pack of 10
Estimated Shipment
03-09-2021
Add to basket
Description
Description
  • Created from Kynar polypropylene
  • Ideal for preventing backflow of fluid
Specifications
Specifications
Natural
Kynar
1/8 x 3/8 in.