Indeholder
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Endrin aldehyde | 7421-93-4 | 1000μg/mL |
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Endrin aldehyde |
| Emballage | 2mL amber prescored glass ampule |
| CAS | 7421-93-4, 67-56-1 |
| CAS min % | 0.1, 99.9 |
| Koncentration eller sammensætningsnoter | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| CAS Max % | 0.1, 99.9 |
| Molekylær formel | C12H8Cl6O |
| Til brug med (udstyr) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| InChI nøgle | HCTWZIFNBBCVGM-YJTREVOLSA-N |
| SMIL | C1C(C2C3C1C4C5(C2(C(C3(C45Cl)Cl)(Cl)Cl)Cl)Cl)C=O |
| Vis mere |
Product intended for laboratory use only