Få mere at vide
Endosulfan-II-1,1,5,5-d4, CDN
Artikelnummer. 30321472
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321472 0.01 g
30319262 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321472 Leverandør CDN Leverandørnr. CDND31290.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale beta-Endosulfan D4
CAS 203716-99-8
Molekylær formel C9 D4 H2 Cl6 O3 S
Mængde 0.01 g
Synonym 6,9-Methano-2,4,3-benzodioxathiepin-1,5-d2, 6,7,8,9,10,10-hexachloro-1,5,5 A,6,9,9 A-hexahydro-1,5-d2-, 3-oxide, (3α,5 Aα,6β,9β,9 Aα)- (9 CI), (3α,5 Aα,6β,9β,9 Aα)-6,7,8,9,10,10-Hexachloro-1,5,5 A,6,9,9 A-hexahydro-1,5-d2-6,9-methano-2,4,3-benzodioxathiepin-1,5-d2 3-oxide, Endosulfan-II-1,1,5,5-d4, beta-Endosulfan D4, β-Endosulfan-d4
InChI formel InChI=1 S/C9H6Cl6O3S/c10-5-6(11)8(13)4-2-18-19(16)17-1-3(4)7(5,12)9(8,14)15/h3-4 H,1-2H2/t3-,4+,7-,8+,19-/i1D2,2D2
SMIL [2 H]C1([2 H])O[S@](=O)OC([2 H])([2 H])[C@@H]2 [C@H]1 [C@]3(Cl)C(=C(Cl)[C@@]2(Cl)C3(Cl)Cl)Cl
IUPAC navn beta-Endosulfan D4
Molekylvægt (g/mol) 410.9498
Formel vægt 407.842 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity