Få mere at vide
MedChemExpress β-Endorphin, human
Artikelnummer. 16422939
Encompass_Preferred
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
16422939 1 mg
1 options
Artikelnummer. 16422939 Leverandør MedChemExpress Leverandørnr. HYP1502
MedChemExpress β-Endorphin, human, a prominent endogenous peptide, existing in the hypophysis cerebri and hypothalamus, is an agonist of opioid receptor, with preferred affinity for μ-opioid receptor and δ-opioid receptor; β-Endorphin, human exhibits antinociception activity.
Tekniske data
Renhed 97.67%
CAS 61214-51-5
Indhold og opbevaring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Til brug med (applikation) Neuroscience-Neuromodulation
Molekylvægt (g/mol) 3464.98 g/mol
Produkttype Reference compound