missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Antibody (SPM255), DyLight 405, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP3-11461V
This item is not returnable.
View return policy
Description
Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Monoclonal antibody specifically detects Endorepellin/Perlecan/Heparan Sulfate Proteoglycan in Human, Mouse, Porcine, Bovine, Fish, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Endorepellin/Perlecan/Heparan Sulfate Proteoglycan | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Pig, Bovine, Fish, Monkey | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SPM255 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1 | |
Murine EHS laminin preparation | |
0.1 mL | |
Cell Biology | |
3339 | |
Store at 4°C in the dark. | |
IgG2a Îş |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Antibody (SPM255), DyLight 405, Novus Biologicals™ > 0.1 mL; DyLight 405
Spot an opportunity for improvement?Share a Content Correction