Beskrivelse
Endophilin A1/SH3GL2 Polyclonal antibody specifically detects Endophilin A1/SH3GL2 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Tekniske data
Tekniske data
| Antigen | Endophilin A1/SH3GL2 |
| Applikationer | Western Blot, Immunocytochemistry, Immunofluorescence |
| Klassifikation | Polyclonal |
| Koncentration | 1mg/mL |
| Konjugeret | Unconjugated |
| Fortynding | Western Blot 1:5000-1:20000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 |
| Formulering | PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal |
| Genadgangsnr. | Q99962 |
| Gene Alias | CNSA2FLJ25015, EEN-B1bA335L15.1 (SH3-domain GRB2-like 2), Endophilin-1, endophilin-A1, FLJ20276, SH3 domain protein 2A, SH3 domain-containing GRB2-like protein 2, SH3D2AEndophilin A1 BAR domain, SH3-domain GRB2-like 2, SH3P4endophilin-1 |
| Gensymboler | SH3GL2 |
| Vis mere |
For Research Use Only