missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Endomorphin-2

Catalog Number p-7057569
Click to view available options
Quantity:
5mg
25mg
This item is not returnable. View return policy
141801-26-5
This item is not returnable. View return policy
141801-26-5

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Content And Storage Store at -20°C
Format Solid
Molecular Weight (g/mol) 571.678
Product Type Endomorphin-2
Quantity 5mg

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'