missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ ENDOG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517329
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
It is not recommended to aliquot this antibody.
The protein encoded by this gene is a nuclear encoded endonuclease that is localized in the mitochondrion. The encoded protein is widely distributed among animals and cleaves DNA at GC tracts. This protein is capable of generating the RNA primers required by DNA polymerase gamma to initiate replication of mitochondrial DNA.
Tekniske data
| ENDOG | |
| Polyclonal | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| O08600, Q14249 | |
| ENDOG | |
| Synthetic peptide corresponding to residues surrounding Cys122 of human Endonuclease G. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| ENDOG | |
| endo G; Endog; endonuclease G; endonuclease G, mitochondrial; mitochondrial endonuclease G | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13804, 2021, 362100 | |
| -20°C | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion