Få mere at vide
endo-alpha-Hydroxy-alpha-phenylbenzeneacetic Acid 8-Azabicyclo[3.2.1]oct-3-yl Ester Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (1R,3r,5S)-8-Azabicyclo(3.2.1)oct-3-yl Hydroxydiphenylacetate Hydrochloride |
| CAS | 63516-30-3 |
| Molekylær formel | C21 H23 N O3 . Cl H |
| Mængde | 500 mg |
| Synonym | Benzeneacetic acid, α-hydroxy-α-phenyl-, (3-endo)-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (1:1), Benzeneacetic acid, α-hydroxy-α-phenyl-, (3-endo)-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (9CI), Benzeneacetic acid, α-hydroxy-α-phenyl-, 8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride, endo- |
| InChI formel | InChI=1S/C21H23NO3.ClH/c23-20(25-19-13-17-11-12-18(14-19)22-17)21(24,15-7-3-1-4-8-15)16-9-5-2-6-10-16;/h1-10,17-19,22,24H,11-14H2;1H/t17-,18+,19+; |
| SMIL | Cl.OC(C(=O)O[C@H]1C[C@H]2CC[C@@H](C1)N2)(c3ccccc3)c4ccccc4 |
| IUPAC navn | [(1S,5R)-8-azabicyclo[3.2.1]octan-3-yl] 2-hydroxy-2,2-diphenylacetate;hydrochloride |
| Molekylvægt (g/mol) | 373.87 |
| Formel vægt | 373.1445 |