Få mere at vide
SHP Steriltechnik AG Endless Cycle Programme
Beskrivelse
- Continuous operation for uninterrupted sterilisation
- Optimised for high-throughput and batch processes
- Energy-efficient thanks to the efficient use of steam and heat
- Reproducible and reliable sterilisation results
Tekniske data
Tekniske data
| Til brug med (udstyr) | Autoclaves |
| Produkttype | Software |