Learn More
SPEX Certiprep™ Extractor for Small Vials
Use this to easily removes the end plug from frozen vials.
Brand:  SPEX Certiprep™ 6756
Product Code. 15360492
  • 8300.00 DKK / Each
Estimated Shipment: 30-06-2025
Explore more special offers
Description
  • This is an end-plug extractor bench-top accessory tool for the SPEX SamplePrep freezer mill small grinding vials (6751, 6761, 6771 and 6781S). 
  • Simply place a vial into the extractor cradle and press down the extractor handle to remove the end plug of the vial.
  • Sample can be retrieved from the vial while it's still cold.
  • The extractor features suction cups that secure the extractor to the benchtop allowing for one-handed operation.
Specifications
Specifications
1
Documents
One moment while we fetch your results.
Product Title
SPEX Certiprep™ Extractor for Small Vials >