Få mere at vide
Tocris Bioscience™ EN6
Beskrivelse
EN6 is an activator of lysosomal vacuolar H+ ATPase (v-ATPase). EN6 covalently binds to the catalytic subunit of v-ATPase leading to increased acidification of lysosomes, inactivation of mTORC1 signaling and activation of autophagy in vivo. EN6 triggers entosis and promotes clearance of TDP43 protein aggregates in vitro.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-[4-Fluoro-3-[(1-oxo-2-propen-1-yl)amino]phenyl]-1-(2-fluorophenyl)-1 H-pyrazole-4-carboxamide |
| CAS | 1808714-73-9 |
| Mængde | 10 mg |
| Mål | H+-ATPase Inhibitors |
| Molekylær formel | C19H14F2N4O2 |
| Renhed | 0.98 |